ALFA43250

43250-02 43250-09 43250-04
AA43250-02EA 47.73 USD
AA43250-02 AA43250-04 AA43250-09
ALFA43250
Applications: In oxidation and photocatalysis
Physical Form: Powder
Sensitivity: Hygroscopic
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR