Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrazine, standard solution, Specpure®, N_2H4 100µg/ml

Supplier: Alfa Aesar
Specpure
42561-AO
AA42561-AOEA 107.74 USD InStock
AA42561-AO
Hydrazine, standard solution, Specpure®, N_2H4 100µg/ml
Standards Environmental Analysis Standards
Liquid

Molecular Formula: N2H4 in 1% CH3CO2H
Formula Weight: 50.06
Physical Form: Liquid
Odor: Ammonia-like
MDL No.: MFCD00011417

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty