Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diyttrium trioxide ≥99.9% (REO, rare earth oxide basis), aerosol Aerosol Refractory Paint

Supplier: Alfa Aesar
Danger
Synonyms: Yttrium (III) oxide Yttrium oxide

40392-CLC 40392-CLC
AA40392-CLCEA 331.66 USD InStock
AA40392-CLC CAAA40392-CLC
Diyttrium trioxide ≥99.9% (REO, rare earth oxide basis), aerosol Aerosol Refractory Paint
Diyttrium trioxide
Provides easy application of uniform, thin, protective high temperature Y╣O╗ coatings. Substrates can be ceramic, graphite or metal. Y╣O╗ offers resistance to molten metals, high temperature lubrication, electrical and thermal insulation.

  • Aerosol Spray Can


Formula: Y₂O₃
MW: 225.81 g/mol
Boiling Pt: –44 °C
Melting Pt: 2410 °C
Density: 5.01 g/cm³ (20 °C)
Flash Pt: –104 °C (–155 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5
UN: 1950
ADR: 2.1,
Order Now
Specification Test Results
Assay Minimum 99.9

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty