Would you like to log in again?
Dropping Bottles, PTFE, Alfa Aesar
Supplier: Alfa Aesar
38106-KT
AA38106-KTEA 91.98 USD InStock
AA38106-KT
Dropping Bottles, PTFE, Alfa Aesar
Bottles Dropping and Dispensing Bottles
PTFE dropping bottles are designed for use with aggressive and valuable reagents with PTFE cap
  • Flexible
  • Inert and leak-free
  • Easy for removal of contents
Ideal for storage and transport of valuable or aggressive materials. The thick walled construction enables the bottle to use for small scale reactions at low pressure.
It features heavy wall construction with super smooth internal finish.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty