Tin, Quant® Test Strips, Thermo Scientific

Supplier: Thermo Scientific
35376-QA
AA35376-QAEA 260.22 USD
AA35376-QA
Tin, Quant® Test Strips, Thermo Scientific
Environmental Test Kits
Odor: Unpleasant
UN No.: 3264
MDL No.: MFCD00148647
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR