ColorpHast® pH Strips, Universal, four squares with different indicators on each strip, 0 to 14 pH range, Thermo Scientific
Supplier: Thermo Scientific
035237-LQ
035237-LS
AA35237-LSEA
262.52
USD
AA35237-LS
AA35237-LQ
ColorpHast® pH Strips, Universal, four squares with different indicators on each strip, 0 to 14 pH range, Thermo Scientific
Test Paper
pH Indicator Papers and Test Strips