ColorpHast® pH Strips, Intermediate, three squares with different indicators on each strip, 7.5 to 14 pH range, Thermo Scientific
Supplier: Thermo Scientific
035233-LS
035233-LQ
AA35233-LSEA
427.99
USD
AA35233-LS
AA35233-LQ
ColorpHast® pH Strips, Intermediate, three squares with different indicators on each strip, 7.5 to 14 pH range, Thermo Scientific
Test Paper
pH Indicator Papers and Test Strips