Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molybdenum, Quant® Test Strips

Supplier: Alfa Aesar
23280-QA 23280-QA
AAA23280-QAEA 258.53 USD InStock
CAAAA23280-QA AAA23280-QA
Molybdenum, Quant® Test Strips
Environmental Test Kits
Reagents & strips to measure molybdenum in aqueous solution.

CAS No.: 5329-14-6
Odor: Odorless
pH: 1.2
UN No.: 2967
MDL No.: MFCD00147655

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty