Hanks' Balanced Salt Solution (1X), without Calcium, without Magnesium, with Phenol Red, Thermo Scientific

Supplier: Thermo Scientific
J67716-K2 J67716-AP
AAJ67716-APEA 83.81 USD
AAJ67716-AP AAJ67716-K2
Hanks' Balanced Salt Solution (1X), without Calcium, without Magnesium, with Phenol Red, Thermo Scientific
Buffers Cell Culture Buffers
Physical Form: Liquid
Appearance: Red
MDL No.: MFCD00217418
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR