Earle's Balanced Salts Solution (10X), Thermo Scientific

Supplier: Thermo Scientific
J67634-K2 J67634-AP
AAJ67634-APEA 104.01 USD
AAJ67634-AP AAJ67634-K2
Earle's Balanced Salts Solution (10X), Thermo Scientific
Buffers Cell Culture Buffers
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00217343
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR