Earle's Balanced Salts Solution (10X), without Calcium Chloride, without Magnesium Sulfate, without Phenol Red, Thermo Scientific

Supplier: Thermo Scientific
J67608-AE J67608-K2
AAJ67608-AEEA 44.15 USD
AAJ67608-AE AAJ67608-K2
Earle's Balanced Salts Solution (10X), without Calcium Chloride, without Magnesium Sulfate, without Phenol Red, Thermo Scientific
Buffers Cell Culture Buffers
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00217343
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR