Gey's Balanced Salt Solution (10X) Kit, with Calcium, with Magnesium, with Phenol Red, Thermo Scientific

Supplier: Thermo Scientific
J67602-AP J67602-K2
AAJ67602-K2EA 143.92 USD
AAJ67602-K2 AAJ67602-AP
Gey's Balanced Salt Solution (10X) Kit, with Calcium, with Magnesium, with Phenol Red, Thermo Scientific
Buffers Cell Culture Buffers
Physical Form: Liquid
Appearance: Red
MDL No.: MFCD00283318
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR