Gey's Balanced Salt Solution (10X), without Calcium, without Magnesium, without Phenol Red, Thermo Scientific

Supplier: Thermo Scientific
J67581-K2 J67581-AP
AAJ67581-K2EA 138.34 USD
AAJ67581-K2 AAJ67581-AP
Gey's Balanced Salt Solution (10X), without Calcium, without Magnesium, without Phenol Red, Thermo Scientific
Buffers Cell Culture Buffers
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00283318
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR