Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bleomycin sulfate (mixture), Cell Culture Grade

Supplier: Alfa Aesar
Danger

J67560-SDL
AAJ67560-SDLEA 700.22 USD InStock
AAJ67560-SDL
Bleomycin sulfate (mixture), Cell Culture Grade
Bleomycin sulfate (mixture)
Bleomycin sulfate binds to DNA, and can inhibit DNA synthesis and causes scission of DNA.

  • White powder

Bleomycin sulfate cleaves DNA, requires binding to oxygen and a metal ion, such as copper or iron. Able to cleave RNA, but at less and more selective degree. Also reported to induce and regulate apoptosis in a variety of cells, and inhibit tumor angiogenesis.

Soluble in water or DMSO

Caution: Store in cool place. Keep container tightly closed in a dry and well-ventilated place.


Formula: C₅₅H₈₄N₁₇O₂₁S₃·xH₂SO₄
MW: 1512,6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070310
CAS Number: 9041-93-4
EINECS: 232-925-2
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty