Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphate-buffered saline (1X), pH 7.4, low endotoxin

Supplier: Alfa Aesar
J67507-AE J67507-AK
AAJ67507-AEEA 50.02 USD InStock
AAJ67507-AE AAJ67507-AK
Phosphate-buffered saline (1X), pH 7.4, low endotoxin
Buffers Cell Culture Buffers
Liquid

Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00131855

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty