QNZ ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: 6-Amino-4-[2-(4-phenoxyphenyl)ethylamino]quinazolineN4-(4-Phenoxyphenethyl)quinazoline-4,6-diamine

J67487-MCR J67487-EXD J67487-LB0
AAJ67487-MCREA 71.82 USD
AAJ67487-MCR AAJ67487-LB0 AAJ67487-EXD
QNZ ≥98%
QNZ

Solid

Formula: C₂₂H₂₀N₄O
MW: 356.429 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06411436
CAS Number: 545380-34-5

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR