Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetate, 1M buffer soln., pH, 4.0, low endotoxin

Supplier: Alfa Aesar
J67486-AK J67486-AE
AAJ67486-AKEA 85.46 USD InStock
AAJ67486-AK AAJ67486-AE
Acetate, 1M buffer soln., pH, 4.0, low endotoxin
Buffers Cell Culture Buffers
Liquid

CAS No.: 127-09-3
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00012459

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty