Dihydrosphingosine-1-phosphate, D-erythro ≥98%

J67482-MCR
AAJ67482-MCREA 340.4 USD
AAJ67482-MCR
Dihydrosphingosine-1-phosphate, D-erythro ≥98%
Dihydrosphingosine-1-phosphate, D-erythro

Solid

Formula: C₁₈H₄₀NO₅P
MW: 381.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00871363
CAS Number: 28508-07-8

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR