Mirin 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
cis-5-(4-Hydroxybenzylidene)-2-imino-4-thiazolidinone
J67462-MF
J67462-MA
J67462-LB0
AAJ67462-MFEA
595.46
USD
AAJ67462-MF
AAJ67462-LB0
AAJ67462-MA
Mirin 95%
Mirin
Solid
Formula:
C₁₀H₈N₂O₂S MW: 220.246 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD05885480 CAS Number: 299953-00-7 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...