Gold 99% supplied in 0.1 mM PBS, nanoparticles 250 nm reactant-free
Supplier: Thermo Scientific Chemicals
J67457-AC
J67457-AE
AAJ67457-ACEA
145.02
USD
AAJ67457-AC
AAJ67457-AE
Gold 99% supplied in 0.1 mM PBS, nanoparticles 250 nm reactant-free
Gold
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications
Liquid
Formula:
Au MW: 196.97 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Specification Test Results
Assay Minimum | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...