SQ 22536 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 9-(Tetrahydrofuran-2-yl)-9H-purin-6-amine9-(Tetrahydro-2-furyl)adenine9-(Tetrahydro-2-furanyl)-9H-purin-6-amine

J67455-MA J67455-MB J67455-LB0
AAJ67455-LB0EA 144.04 USD
AAJ67455-LB0 AAJ67455-MB AAJ67455-MA
SQ 22536 95%
SQ 22536

Solid

Formula: C₉H₁₁N₅O
MW: 205.221 g/mol
Melting Pt: 160…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00210216
CAS Number: 17318-31-9

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR