Farnesyl thiotriazole 98%

Warning

J67454-MA
AAJ67454-MAEA 168.26 USD
AAJ67454-MA
Farnesyl thiotriazole 98%
Farnesyl thiotriazole

Oil

Formula: C₁₇H₂₆N₃S
MW: 305.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270863
CAS Number: 156604-45-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR