N-Linoleoylglycine 98%

J67453-MA J67453-LB0
AAJ67453-LB0EA 160.55 USD
AAJ67453-LB0 AAJ67453-MA
N-Linoleoylglycine 98%
N-Linoleoylglycine

Solid


Caution: Storage Temperature: –30°C to –10°C

Formula: C₂₀H₃₅NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11977729
CAS Number: 2764-03-6

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR