Gold 99% supplied in 0.1 mM PBS, nanoparticles 200 nm reactant-free

J67446-AC J67446-AE
AAJ67446-AEEA 431.19 USD
AAJ67446-AE AAJ67446-AC
Gold 99% supplied in 0.1 mM PBS, nanoparticles 200 nm reactant-free
Gold

Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications


Liquid

Formula: Au
MW: 196.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR