E6 Berbamine 98%

Warning

J67434-MA
AAJ67434-MAEA 233.82 USD
AAJ67434-MA
E6 Berbamine 98%
E6 Berbamine

Yellow Solid


Caution: Storage Temperature: –30°C to –10°C

Formula: C₄₄H₄₃N₃O₉
MW: 757.84 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00676930
CAS Number: 73885-53-7

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR