Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilized, nanoparticles 50 nm OD1 (Abs. 535 nm)
Supplier: Thermo Scientific Chemicals
J67433-AC
J67433-AE
AAJ67433-ACEA
92.9
USD
AAJ67433-AC
AAJ67433-AE
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilized, nanoparticles 50 nm OD1 (Abs. 535 nm)
Gold
For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.
Liquid
Formula:
Au MW: 196.97 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...