Gold 99% supplied in 0.1 mM PBS, nanoparticles 50 nm reactant-free OD1 (Abs. 535 nm)
Supplier: Thermo Scientific Chemicals
J67432-AE
J67432-AC
AAJ67432-ACEA
111.56
USD
AAJ67432-AC
AAJ67432-AE
Gold 99% supplied in 0.1 mM PBS, nanoparticles 50 nm reactant-free OD1 (Abs. 535 nm)
Gold
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications
Liquid
Formula:
Au MW: 196.97 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Specification Test Results
Assay Minimum | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...