15-Deoxy-δ-12,14-prostaglandin J2 ≥97%
|
Danger
|
J67427-MCR
AAJ67427-MCREA
248.42
USD
AAJ67427-MCR
15-Deoxy-δ-12,14-prostaglandin J2 ≥97%
15-Deoxy-delta-12,14-prostaglandin J2
Oil
Formula:
C₂₀H₂₈O₃ MW: 316.441 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00065806 CAS Number: 87893-55-8 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...