D-erythro MAPP 98%

J67412-LB0 J67412-MB
AAJ67412-MBEA 498.31 USD
AAJ67412-MB AAJ67412-LB0
D-erythro MAPP 98%
D-erythro MAPP

Solid

Formula: C₂₃H₃₉NO₂
MW: 361.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01076527
CAS Number: 143492-38-0

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR