HEPES Buffer, 0.2M Buffer Soln., pH 7.0, Low Endotoxin, Thermo Scientific

Supplier: Thermo Scientific
J67405-AE J67405-AK
AAJ67405-AKEA 102.82 USD
AAJ67405-AK AAJ67405-AE
HEPES Buffer, 0.2M Buffer Soln., pH 7.0, Low Endotoxin, Thermo Scientific
Buffers Cell Culture Buffers
Liquid

CAS No.: 7365-45-9
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00006158
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR