Gold 95% supplied in 0.1 mM PBS, nanoparticles 300 nm

J67400-AC J67400-AE
AAJ67400-AEEA 684.15 USD
AAJ67400-AE AAJ67400-AC
Gold 95% supplied in 0.1 mM PBS, nanoparticles 300 nm
Gold

For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM


Liquid

Formula: Au
MW: 196.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR