Sodium Phosphate, 0.2M Buffer Soln., pH 7.2, Low Endotoxin, Thermo Scientific

Supplier: Thermo Scientific
J67394-AE J67394-AK
AAJ67394-AEEA 74.87 USD
AAJ67394-AE AAJ67394-AK
Sodium Phosphate, 0.2M Buffer Soln., pH 7.2, Low Endotoxin, Thermo Scientific
Buffers Cell Culture Buffers
Liquid

Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00146206
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR