scriptaid ≥97%

Synonyms: 6-(1,3-Dioxo-1H-benzo[de]isoquinolin-2(3H)-yl)-N-hydroxyhexanamide

J67379-MCR
AAJ67379-MCREA 130.92 USD
AAJ67379-MCR
scriptaid ≥97%
scriptaid

Solid

Formula: C₁₈H₁₈N₂O₄
MW: 326.352 g/mol
Melting Pt: 160…161 °C
Storage Temperature: Freezer
MDL Number: MFCD00386477
CAS Number: 287383-59-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR