Acetate, 1M Buffer Soln., pH, 3.0, Low Endotoxin, Thermo Scientific

Supplier: Thermo Scientific
J67373-AK J67373-AE
AAJ67373-AEEA 47.34 USD
AAJ67373-AE AAJ67373-AK
Acetate, 1M Buffer Soln., pH, 3.0, Low Endotoxin, Thermo Scientific
Buffers Cell Culture Buffers
Liquid

CAS No.: 127-09-3
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00012459
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR