Nullscript ≥97%
Synonyms:
4-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-N-hydroxybutanamide
J67363-MCR
AAJ67363-MCREA
136.1
USD
AAJ67363-MCR
Nullscript ≥97%
Nullscript
White solid
- Light Sensitive
Solid
Purity: 97%
CAS No.: 300816-11-9
Molecular Formula: C16H14N2O4
Formula Weight: 298.30
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: White
Melting Point: 171-172°
MDL No.: MFCD00405081
Formula:
C₁₆H₁₄N₂O₄ MW: 298.30 g/mol Melting Pt: 171…172 °C Storage Temperature: Freezer |
MDL Number:
MFCD00405081 CAS Number: 300816-11-9 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...