Manoalide ≥98%
Supplier: Thermo Scientific Chemicals
J67341-MCR
AAJ67341-MCREA
375.38
USD
AAJ67341-MCR
Manoalide ≥98%
Manoalide
Solid
Formula:
C₂₅H₃₆O₅ MW: 416.558 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00153826 CAS Number: 75088-80-1 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...