4-Phenylchalcone oxide ≥99%

J67334-FPL
AAJ67334-FPLEA 166.83 USD
AAJ67334-FPL
4-Phenylchalcone oxide ≥99%
4-Phenylchalcone oxide

Solid

Formula: C₂₁H₁₆O₂
MW: 300.357 g/mol
Melting Pt: 257…259 °C
Storage Temperature: Freezer
MDL Number: MFCD00211155
CAS Number: 82389-34-2

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR