Cp 31398 Dihydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N'-[2-[2-(4-Methoxyphenyl)ethenyl]-4-quinazolinyl]-N,N-dimethyl-1,3-propanediamine dihydrochloride

J67317-MA
AAJ67317-MAEA 249.56 USD
AAJ67317-MA
Cp 31398 Dihydrochloride 98%
Cp 31398 Dihydrochloride

Solid

Formula: C₂₂H₂₈Cl₂N₄O
MW: 435.393 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11519956
CAS Number: 259199-65-0
EINECS: 683-708-8

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR