SB-366791 ≥99%

Synonyms: 4-Chloro-N-(3-methoxyphenyl)cinnamideN-(3-Methoxyphenyl)-4-chlorocinnamide

J67312-LB0
AAJ67312-LB0EA 130.92 USD
AAJ67312-LB0
SB-366791 ≥99%
SB-366791

Solid

Formula: C₁₆H₁₄ClNO₂
MW: 287.743 g/mol
Melting Pt: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD01033818
CAS Number: 472981-92-3

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR