Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilized, nanoparticles 60 nm OD1 (Abs. 540 nm)
Supplier: Thermo Scientific Chemicals
J67306-AC
J67306-AE
AAJ67306-AEEA
318.07
USD
AAJ67306-AE
AAJ67306-AC
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilized, nanoparticles 60 nm OD1 (Abs. 540 nm)
Gold
For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.
Liquid
Formula:
Au MW: 196.97 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...