Ar231453 ≥98%

Warning

J67299-LB0
AAJ67299-LB0EA 175.3 USD
AAJ67299-LB0
Ar231453 ≥98%
Ar231453

Solid

Formula: C₂₁H₂₄FN₇O₅S
MW: 505.525 g/mol
Melting Pt: 174…176 °C
Storage Temperature: Freezer
MDL Number: MFCD14706714
CAS Number: 733750-99-7

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR