Potassium Phosphate, 0.2M Buffer Soln., pH 7.5, Low Endotoxin, Thermo Scientific

Supplier: Thermo Scientific
J67297-AE J67297-AK
AAJ67297-AKEA 64.52 USD
AAJ67297-AK AAJ67297-AE
Potassium Phosphate, 0.2M Buffer Soln., pH 7.5, Low Endotoxin, Thermo Scientific
Buffers Cell Culture Buffers
Liquid

CAS No.: 7778-77-0
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00011401
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR