Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bicine, 0.2M buffer soln., pH 8.5, low endotoxin

Supplier: Alfa Aesar
J67296-AE J67296-AK
AAJ67296-AEEA 51.51 USD InStock
AAJ67296-AE AAJ67296-AK
Bicine, 0.2M buffer soln., pH 8.5, low endotoxin
Buffers Cell Culture Buffers
Liquid

CAS No.: 150-25-4
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00004295

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty