N-Arachidonyl-2-chloroethylamide 96%

J67294-MA
AAJ67294-MAEA 134.81 USD
AAJ67294-MA
N-Arachidonyl-2-chloroethylamide 96%
N-Arachidonyl-2-chloroethylamide

Oil

Formula: C₂₂H₃₈ClNO₂
MW: 384.001 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02683581
CAS Number: 220556-69-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR