Cantharidic acid 98%

Warning

J67282-MA
AAJ67282-MAEA 139.9 USD
AAJ67282-MA
Cantharidic acid 98%
Cantharidic acid

White solid

Formula: C₁₀H₁₄O₅
Melting Pt: 216…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00047137
CAS Number: 28874-45-5

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR