Ilimaquinone ≥98%

J67278-EXG
AAJ67278-EXGEA 256.77 USD
AAJ67278-EXG
Ilimaquinone ≥98%
Ilimaquinone

Oil

Formula: C₂₂H₃₀O₄
MW: 358.478 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22200891
CAS Number: 71678-03-0

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR