P32/98 98%

Supplier: Thermo Scientific Chemicals
Warning

J67268-MA
AAJ67268-MAEA 258.71 USD
AAJ67268-MA
P32/98 98%
P32/98

Solid

Formula: C₁₃H₂₂N₂O₅S
MW: 318.388 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22741399
CAS Number: 136259-20-6

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR