N-Arachidonoyldopamine 98%

J67250-LB0 J67250-MB
AAJ67250-LB0EA 118.31 USD
AAJ67250-LB0 AAJ67250-MB
N-Arachidonoyldopamine 98%
N-Arachidonoyldopamine

Viscous oil

Formula: C₂₈H₄₁NO₃
MW: 439.64 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03412031
CAS Number: 199875-69-9

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR