Src inhibitor-1 ≥98%
|
Warning
|
Synonyms:
6,7-Dimethoxy-N-(4-phenoxyphenyl)quinazolin-4-amine
J67242-LB0
AAJ67242-LB0EA
172.46
USD
AAJ67242-LB0
Src inhibitor-1 ≥98%
Src inhibitor-1
White solid
Solid
Purity: 98%
CAS No.: 179248-59-0
Molecular Formula: C22H19N3O3
Formula Weight: 373.41
Physical Form: Solid
Appearance: White
MDL No.: MFCD01815300
Formula:
C₂₂H₁₉N₃O₃ MW: 373.41 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01815300 CAS Number: 179248-59-0 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...