Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Farnesylthioacetic acid ≥98%

Supplier: Alfa Aesar

J67240-LB0
AAJ67240-LB0EA 132.65 USD InStock
AAJ67240-LB0
Farnesylthioacetic acid ≥98%
Farnesylthioacetic acid
Yellow oil

  • Light Sensitive

Oil

Purity: 98%
CAS No.: 135784-48-4
Molecular Formula: C17H28O2S
Formula Weight: 296.47
Storage Temperature: -30°C to -10°C
Physical Form: Oil
Sensitivity: Light Sensitive
Appearance: Yellow
MDL No.: MFCD00216146


Formula: C₁₇H₂₈O₂S
MW: 296,47 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00216146
CAS Number: 135784-48-4
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty